missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic powder, -20 mesh, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1010118

 View more versions of this product

Catalog No. AA1010118



Specifications

Specifications

Arsenic powder
7440-38-2
As
UN1558
arsen, arsenic, inorganic, arsenic-75, arsenicals, black, colloidal, compounds, gray, grey, metallic
[As]
Odorless
5359596
Powder
Black to Silver
50 g
(metals basis)
MFCD00085309
−20 Mesh
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
74.922
CHEBI:27563
99%

Chemical Identifiers

7440-38-2
74.922
RQNWIZPPADIBDY-UHFFFAOYSA-N
5359596
arsenic
As
MFCD00085309
arsen, arsenic, inorganic, arsenic-75, arsenicals, black, colloidal, compounds, gray, grey, metallic
CHEBI:27563
[As]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC

EINECSNumber : 213-148-6

RTECSNumber : CG0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents