missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.99999+% (metals basis)

Manufacturer:  Alfa Aesar™ 1304414

 View more versions of this product

Catalog No. AA1304414



Description

Description

  • Packaged under argon
Specifications

Specifications

Arsenic polycrystalline lump
Gray
2 to 8mm (0.08 to 0.3 in.)
Odorless
As
UN1558
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
74.92
Puratronic™
Air Sensitive
25g
7440-38-2
(metals basis)
MFCD00085309
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
[As]
74.922
CHEBI:27563
≥99.99999%

Chemical Identifiers

7440-38-2
74.922
RQNWIZPPADIBDY-UHFFFAOYSA-N
5359596
arsenic
As
MFCD00085309
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
CHEBI:27563
[As]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC

RTECSNumber : CG0525000

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents