missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01304409

 View more versions of this product

Catalog No. AA1304409



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Gray
2 to 8mm (0.08 to 0.3 in.)
(metals basis)
MFCD00085309
10 g
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
74.92
Puratronic™
Air Sensitive
Arsenic polycrystalline lump
7440-38-2
As
UN1558
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
[As]
74.92
CHEBI:27563
≥99.99999%
Odorless

Chemical Identifiers

7440-38-2
74.92
RQNWIZPPADIBDY-UHFFFAOYSA-N
5359596
arsenic
As
MFCD00085309
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
CHEBI:27563
[As]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC

EINECSNumber : 213-148-6

RTECSNumber : CG0525000

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only