missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1337503

 View more versions of this product

Catalog No. AA1337503



Specifications

Specifications

Arsenic
99.9999% (Metals basis)
MFCD00085309
arsen
[As]
74.922
CHEBI:27563
Odorless
1g
7440-38-2
As
Gray lump
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
Polycrystalline Lump, 2-8mm (0.08-0.3 in.)
Gray

Chemical Identifiers

7440-38-2
74.922
RQNWIZPPADIBDY-UHFFFAOYSA-N
5359596
arsenic
As
MFCD00085309
arsen
CHEBI:27563
[As]
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 213-148-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents