missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1337509

 View more versions of this product

Catalog No. AA1337509



Specifications

Specifications

Arsenic
99.9999% (Metals basis)
MFCD00085309
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
[As]
74.922
CHEBI:27563
Odorless
10g
7440-38-2
As
Gray lump
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
Polycrystalline Lump, 2-8mm (0.08-0.3 in.)
Gray

Chemical Identifiers

7440-38-2
74.922
RQNWIZPPADIBDY-UHFFFAOYSA-N
5359596
arsenic
As
MFCD00085309
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
CHEBI:27563
[As]
Safety and Handling

Safety and Handling

H301-H331-H400-H410; Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 213-148-6

Documents