missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13836AD

 View more versions of this product

Catalog No. AA13836AD



Specifications

Specifications

Arsenic
Clear liquid
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenic
5359596
Liquid
Colorless
7440-38-2,7440-38-2,7732-18-5
As
Nitric acid: 5.0%; Arsenic: 0.1%; Water: 94.9%
[As]
74.922
CHEBI:27563
23 hPa (17mm Hg) at 20°C
50mL

Chemical Identifiers

7440-38-2
74.922
RQNWIZPPADIBDY-UHFFFAOYSA-N
CHEBI:27563
[As]
As
MFCD00085309
5359596
arsenic
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents