Learn More
Arsenic(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals
$238.32 - $238.32
Chemical Identifiers
| CAS | 12044-54-1 |
|---|---|
| Molecular Formula | As2Te3 |
| Molecular Weight (g/mol) | 532.643 |
| MDL Number | MFCD00014167 |
| InChI Key | GTIUFDICMGTSPM-UHFFFAOYSA-N |
| Synonym | arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride |
| PubChem CID | 15770922 |
| SMILES | [As](=[Te])[Te][As]=[Te] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12044-54-1 | |
| 532.643 | |
| GTIUFDICMGTSPM-UHFFFAOYSA-N | |
| 15770922 |
| As2Te3 | |
| MFCD00014167 | |
| arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride | |
| [As](=[Te])[Te][As]=[Te] |
Specifications
| 381°C | |
| Powder | |
| As2Te3 | |
| UN1557 | |
| GTIUFDICMGTSPM-UHFFFAOYSA-N | |
| 532.643 | |
| 532.64 | |
| Odorless | |
| Arsenic(III) telluride |
| 12044-54-1 | |
| 5 g | |
| MFCD00014167 | |
| arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride | |
| [As](=[Te])[Te][As]=[Te] | |
| 15770922 | |
| 99.999% | |
| 6.50 g/mL | |
| (metals basis) |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 234-955-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only