missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic(III) sulfide, Yellow fused pieces, 99.99% (Metals basis), Alfa Aesar™

Chemical Identifiers

CAS 1303-33-9
Physical Form Fused Pieces
Assay Percent Range 99.99% (Metals basis)
Molecular Formula As2S3
Formula Weight 246.03
MDL Number MFCD00003435
Chemical Name or Material Arsenic(III) sulfide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4334006 SDS Alfa Aesar
4334006
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4334014 SDS Alfa Aesar
4334014
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1303-33-9
99.99% (Metals basis)
246.03
Arsenic(III) sulfide
Fused Pieces
As2S3
MFCD00003435

Specifications

1303-33-9
99.99% (Metals basis)
246.03
3.43g/cm3
Yellow
Arsenic(III) sulfide
Fused Pieces
As2S3
MFCD00003435
Like rotten eggs
5g

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

H301-H331-H400-H410: Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 215-117-4