Please login to your online account to display your discounted pricing

Arsenic(III) sulfide, Powder, 99.99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Arsenic(III) sulfide
CAS 1303-33-9
Physical Form Powder
Assay Percent Range 99.99% (Metals basis)
Molecular Formula As2S3
Formula Weight 246.03
MDL Number MFCD00003435
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4552214

 
alfa aesar
4552214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.03
Quantity 25g
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Arsenic(III) sulfide
EINECS Number 215-117-4
Physical Form Powder
Assay Percent Range 99.99% (Metals basis)
Molecular Formula As2S3
MDL Number MFCD00003435
CAS 1303-33-9
Health Hazard 1 H301-H331-H400-H410: Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Health Hazard 2 When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
Safety and Handling

H301-H331-H400-H410: Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.