missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(III) sulfide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4561606

 View more versions of this product

Catalog No. AA4561606



Specifications

Specifications

Arsenic (III) sulfide
(metals basis)
MFCD00003435
14,806
Powder
3.43g/mL
707°C
5g
1303-33-9
As2S3
UN1557
246.03
99.9%
Like rotten eggs
300°C

Chemical Identifiers

1303-33-9
As2S3
MFCD00003435
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P273-P270-P309-P310-P302+P352-P501a
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

RTECSNumber : CG2638000

TSCA : TSCA

Documents