Learn More
Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04561614
Description
Arsenic(III) sulfide is used in the treatment of acute promyelocytic leukemia. It is an inorganic photoresist used in the fabrication of photonic crystals and 3-D nanostructures. It is also used as an acousto-optic material, pigment, tanning agent and chalcogenide glass for infrared optics. Further, it is used as an analytical test for presence of dissimilatory arsenic-reducing bacteria (DARB).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
707°C | |
300°C | |
Powder | |
(metals basis) | |
MFCD00003435 | |
14,806 | |
CUGMJFZCCDSABL-UHFFFAOYSA-N | |
diarsenic(3+) trisulfanediide | |
246.03 | |
Like rotten eggs |
Arsenic (III) sulfide | |
1303-33-9 | |
25 g | |
As2S3 | |
UN1557 | |
Insoluble in water. | |
[S--].[S--].[S--].[As+3].[As+3] | |
246.02 | |
99.9% | |
3.43 g/mL |
Chemical Identifiers
1303-33-9 | |
246.02 | |
CUGMJFZCCDSABL-UHFFFAOYSA-N | |
[S--].[S--].[S--].[As+3].[As+3] |
As2S3 | |
MFCD00003435 | |
diarsenic(3+) trisulfanediide |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber : 215-117-4
RTECSNumber : CG2638000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only