missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(III) selenide, 99.999% (metals basis)

For vacuum deposition

Manufacturer:  Alfa Aesar™ 1284902

 View more versions of this product

Catalog No. AA1284902



Specifications

Specifications

Arsenic(III) selenide
(metals basis)
MFCD00014166
14,805
WBFMCDAQUDITAS-UHFFFAOYSA-N
386.756
386.72
99.999%
Odorless
260°C
1303-36-2
As2Se3
UN3077
arsenic triselenide, diarsenic triselenide, arsenic selenide, arsenous selenide, arsenic sesquiselenide, arsenic selenide, unii-xws0w0gb8g, diarsenic triselenide asse3, arsenic iii selenide, xws0w0gb8g
[As](=[Se])[Se][As]=[Se]
14772
Fused Pieces
4.75g/mL
1 to 6mm
0.5g

Chemical Identifiers

1303-36-2
386.756
WBFMCDAQUDITAS-UHFFFAOYSA-N
14772
As2Se3
MFCD00014166
arsenic triselenide, diarsenic triselenide, arsenic selenide, arsenous selenide, arsenic sesquiselenide, arsenic selenide, unii-xws0w0gb8g, diarsenic triselenide asse3, arsenic iii selenide, xws0w0gb8g
[As](=[Se])[Se][As]=[Se]
Safety and Handling

Safety and Handling

GHS H Statement
H330
Fatal if inhaled.

GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : CG2285000

TSCA : TSCA

Documents