missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic(III) selenide, 99.999% (metals basis), Alfa Aesar™

Used as a vacuum deposition, chalcogenide glass for infrared optics, as it transmits light with wavelengths between 870 nm and 17.2 µm.

Manufacturer:  Alfa Aesar 4374806

 View more versions of this product

Catalog No. AA4374806



Specifications

Specifications

Arsenic(III) selenide
4.75
1303-36-2
MFCD00014166
arsenic iii selenide
[As](=[Se])[Se][As]=[Se]
14772
99.999%
260°C
Glass bottle
5g
As2Se3
UN3077
WBFMCDAQUDITAS-UHFFFAOYSA-N
386.756
386.72
−160 Mesh Powder

Chemical Identifiers

1303-36-2
386.756
WBFMCDAQUDITAS-UHFFFAOYSA-N
14772
As2Se3
MFCD00014166
arsenic iii selenide
[As](=[Se])[Se][As]=[Se]
Safety and Handling

Safety and Handling

TSCA : Yes

Documents