Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(III) selenide, 99.999% (metals basis)

For vacuum deposition

Chemical Identifiers

Chemical Name or Material Arsenic(III) selenide
CAS 1303-36-2
Formula Weight 386.72
MDL Number MFCD00014166
UN Number UN3077
Merck Index 14,805
Synonym Arsenic triselenide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1284906

  SDS
alfa aesar™
1284906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1284902

  SDS
alfa aesar™
1284902
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1284914

  SDS
alfa aesar™
1284914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 386.72
Chemical Name or Material Arsenic(III) selenide
TSCA TSCA
Odor Odorless
Particle Size 1 to 6mm
Physical Form Fused Pieces
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00014166
CAS 1303-36-2
UN Number UN3077
Merck Index 14,805
Synonym Arsenic triselenide
Melting Point 260°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.75g/mL
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number CG2285000
Safety and Handling

Danger

GHS P Statement
P260-P284-P304+P340-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear respiratory protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330
Fatal if inhaled.