missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb 20ppm max

Manufacturer:  Alfa Aesar™ 1117318

 View more versions of this product

Catalog No. AA1117318



Specifications

Specifications

Arsenic(III) oxide
(metals basis excluding Sb), Sb 20ppm max.
UN1561
Soluble in hot water,dilute acids and alkalies. Insoluble in alcohol,chloroform,and ether,.
Powder
Puratronic™
Odorless
312.3°C
1327-53-3
MFCD00003433
14,804
197.84
99.996%
3.74 g/mL
465°C (sublimation)
50g

Chemical Identifiers

1327-53-3
MFCD00003433
Safety and Handling

Safety and Handling

GHS H Statement
H300-H350-H314-H318
Fatal if swallowed.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300-H314-H335-H350-H371

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC TRIOXIDE

EINECSNumber : 215-481-4

RTECSNumber : CG3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents