missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(III) oxide, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 43488A1

 View more versions of this product

Catalog No. AA43488A1



Specifications

Specifications

Arsenic (III) oxide
(metals basis)
MFCD00003433
14,804
Slightly soluble in cold water; Soluble in boiling water,HCl,alkali hydroxide; Insoluble in alcohol,chloroform,and etherSoluble in dilute acids and alkalies; Insoluble in organic solvents; Slightly soluble in water.
-100 Mesh Powder
3.74 g/mL
Odorless
312.3°C
1327-53-3
As2O3
UN1561
Arsenic trioxide
197.84
99.5%
−100 Mesh
465°C (sublimation)
1kg

Chemical Identifiers

1327-53-3
As2O3
MFCD00003433
Arsenic trioxide
Safety and Handling

Safety and Handling

GHS H Statement
H300-H350-H314-H318
Fatal if swallowed.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300-H314-H335-H350-H371

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC TRIOXIDE

EINECSNumber : 215-481-4

RTECSNumber : CG3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents