missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic(III) iodide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04506709

 View more versions of this product

Catalog No. AA4506709



Description

Description

Arsenic(III) iodide is mainly used to treat rheumatism, arthritis, malaria, trypanosome infections, tuberculosis, and diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

424°C
140°C
Pieces
AsI3
UN1557
arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid
IKIBSPLDJGAHPX-UHFFFAOYSA-N
triiodoarsane
24575
98%
Moisture sensitive
Arsenic (III) iodide
7784-45-4
10 g
MFCD00014170
14,803
Soluble in alcohol,ether,Carbon disulfide,water(6g/100 m).
[As](I)(I)I
455.635
455.64
Pungent
4.39 g/mL

Chemical Identifiers

7784-45-4
455.635
IKIBSPLDJGAHPX-UHFFFAOYSA-N
24575
[As](I)(I)I
AsI3
MFCD00014170
arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid
triiodoarsane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

EINECSNumber : 232-068-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only