Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(III) chloride, Reagent Grade

Chemical Identifiers

Chemical Name or Material Arsenic(III) chloride
CAS 7784-34-1
Purity Grade Reagent Grade
Formula Weight 181.27
MDL Number MFCD00014165
UN Number UN1560
Merck Index 14,801
Synonym Arsenic trichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3149918

  SDS
alfa aesar™
3149918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3149930

  SDS
alfa aesar™
3149930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.27
Chemical Name or Material Arsenic(III) chloride
TSCA TSCA
Odor Acrid
Purity Grade Reagent Grade
Physical Form Liquid
Sensitivity Moisture Sensitive
MDL Number MFCD00014165
CAS 7784-34-1
UN Number UN1560
Merck Index 14,801
Synonym Arsenic trichloride
Boiling Point 130.2°C
Melting Point −16°C
Quantity 50g
Solubility Information Decomposes in water with formation of As(OH)3 and HCl; Increasing dilutions in water result in As2O3 precipitation
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H331-H350-H314
Fatal if swallowed.
Toxic if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P273-P201-P309-P310-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.163g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ARSENIC TRICHLORIDE
RTECS Number CG1750000
Safety and Handling

Danger

GHS P Statement
P273-P201-P309-P310-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H331-H350-H314
Fatal if swallowed.
Toxic if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.