Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(III) chloride, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Arsenic(III) chloride
CAS 7784-34-1
Formula Weight 181.27
MDL Number MFCD00014165
UN Number UN1560
Merck Index 14,801
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1168106

 
alfa aesar™
1168106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1168114

 
alfa aesar™
1168114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1168122

 
alfa aesar™
1168122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.27
Chemical Name or Material Arsenic(III) chloride
TSCA TSCA
Odor Acrid
Physical Form Liquid
Assay 99.999%
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
MDL Number MFCD00014165
CAS 7784-34-1
UN Number UN1560
Merck Index 14,801
Boiling Point 130.2°C
Color Colorless to yellow-brown
Melting Point −16°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H331-H350-H314
Fatal if swallowed.
Toxic if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P273-P201-P309-P310-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.163g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ARSENIC TRICHLORIDE
RTECS Number CG1750000
Safety and Handling

Danger

GHS P Statement
P273-P201-P309-P310-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H331-H350-H314
Fatal if swallowed.
Toxic if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.