Learn More
Arsenic(II) sulfide, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01728414
Description
Arsenic(II) sulfide is used as an primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
565°C | |
∼307°C | |
Powder | |
AsH3S | |
UN1557 | |
Insoluble in water. Soluble in acids and alkalis. | |
S[AsH2] | |
110.01 | |
90% | |
Odorless |
Arsenic(II) sulfide | |
12044-79-0 | |
25 g | |
MFCD00049841 | |
arsenic ii sulfide, realgar ass, diarsanedithione | |
BMOFEAKQXGGUPE-UHFFFAOYSA-N | |
arsanethiol | |
213.97 | |
Technical | |
3.5 g/mL |
Chemical Identifiers
12044-79-0 | |
110.01 | |
BMOFEAKQXGGUPE-UHFFFAOYSA-N | |
arsanethiol |
AsH3S | |
MFCD00049841 | |
arsenic ii sulfide, realgar ass, diarsanedithione | |
S[AsH2] |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only