missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Arsenic(II) sulfide, tech. 90%

Manufacturer:  Alfa Aesar™ 1728406

 View more versions of this product

Catalog No. AA1728406



Specifications

Specifications

Arsenic(II) sulfide
MFCD00049841
arsenic ii sulfide, realgar ass, diarsanedithione, arsenic ii sulfide
Powder
Technical
Odorless
~307°C
12044-79-0
UN1557
213.97
90%
3.5g/mL
565°C
5g

Chemical Identifiers

12044-79-0
MFCD00049841
arsenic ii sulfide, realgar ass, diarsanedithione, arsenic ii sulfide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P273-P270-P309-P310-P302+P352-P501a
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

SDS Documents