missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Arsenic ICP Standard, 10,000 ppm As in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PAS10KN100
Specifications
Arsenic ICP Standard | |
Natural LDPE Bottle | |
67.0,30.24,0.67 | |
Colorless | |
As | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
arsenic | |
5359596 | |
1 mL = 10 mg As |
7440-38-2,7697-37-2,7440-38-2,7732-18-5 | |
100 mL | |
70.0,31.48,0.7 | |
10,000ppm As | |
MFCD00085309 | |
[As] | |
74.92 | |
CHEBI:27563 |
Chemical Identifiers
7440-38-2 | |
74.92 | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
CHEBI:27563 | |
[As] |
As | |
MFCD00085309 | |
5359596 | |
arsenic |