missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg As), LabChem™

Manufacturer:  LabChem LC115007

 View more versions of this product

Catalog No. LC115007



Specifications

Specifications

Arsenic AA Standard
1327-53-3,7697-37-2,1303-28-2,7732-18-5
As2O5
UN2031
Liquid
Certified
Soluble in water
1.04mPa.s
-7°C
Colorless
1000ppm (1mL = 1mg As)
95.83,4,0.17
1000ppm ±10ppm (1mL = 1mg As)
229.84
Passes Test
Poly Bottle
Traceable to NIST
1.02g/mL
102°C
125mL

Chemical Identifiers

1327-53-3
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause cancer.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust, mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

Documents
Promotions

Promotions