missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arsenic AA Standard, 1mL = 1mg As (1,000ppm As) in 3% H2SO4, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIAAS1KS100

 View more versions of this product

Catalog No. AAS1KS-100


 

Specifications Description & Specifications

Specifications

Arsenic AA Standard
93.0,5.12,0.12
Liquid
1,000ppm As
Clear colorless
7732-18-5,7664-93-9,1327-53-3
97.0,5.33,0.13
Poly natural
1 mL = 1 mg As
100mL

Chemical Identifiers

7732-18-5,7664-93-9,1327-53-3
97.0,5.33,0.13
Arsenic AA Standard
93.0,5.12,0.12
Liquid