Please login to your online account to display your discounted pricing

Arsenazo III Indicator, Certified, LabChem™

$51.69 - $51.69

Chemical Identifiers

Chemical Name or Material Arsenazo III Indicator
CAS 7732-18-5
CAS Max % 99.8
CAS 1668-00-4
CAS Max % 0.2
Purity Grade Certified Grade
Linear Formula C22H18As2N4O14S2
Formula Weight 776.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC114707

 
lab chem
LC114707
Poly Bottle 125mL Each for $51.69
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 776.37
Chemical Name or Material Arsenazo III Indicator
Quantity 125mL
Physical Form Liquid
Linear Formula C22H18As2N4O14S2
CAS 7732-18-5
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99.8
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 1668-00-4
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Specific Gravity 1g/mL
CAS Max % 0.2
Recommended Storage Room Temperature
Decomposition Information Arsenic and its oxides; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Density 1g/mL
Color Clear, reddish-purple
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature