missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Arotinolol Hydrochloride 98.0+%, TCI America™

$191.48 - $568.55

Chemical Identifiers

CAS 68377-91-3
Molecular Formula C15H22ClN3O2S3
Molecular Weight (g/mol) 407.99
MDL Number MFCD00941421
InChI Key XXDAXBZYUXLDRD-UHFFFAOYNA-N
Synonym 5-[2-[[3-(tert-Butylamino)-2-hydroxypropyl]thio]thiazol-4-yl]thiophene-2-carboxamide Hydrochloride
PubChem CID 155032
IUPAC Name hydrogen 5-(2-{[3-(tert-butylamino)-2-hydroxypropyl]sulfanyl}-1,3-thiazol-4-yl)thiophene-2-carboxamide chloride
SMILES [H+].[Cl-].CC(C)(C)NCC(O)CSC1=NC(=CS1)C1=CC=C(S1)C(N)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A314550MG
View Documents
Tci America
A314550MG
50mg
Each for $191.48
 
A3145250MG
View Documents
Tci America
A3145250MG
250mg
Each for $568.55
 
Description

Description

Specifications

Chemical Identifiers

68377-91-3
407.99
XXDAXBZYUXLDRD-UHFFFAOYNA-N
155032
[H+].[Cl-].CC(C)(C)NCC(O)CSC1=NC(=CS1)C1=CC=C(S1)C(N)=O
C15H22ClN3O2S3
MFCD00941421
5-[2-[[3-(tert-Butylamino)-2-hydroxypropyl]thio]thiazol-4-yl]thiophene-2-carboxamide Hydrochloride
hydrogen 5-(2-{[3-(tert-butylamino)-2-hydroxypropyl]sulfanyl}-1,3-thiazol-4-yl)thiophene-2-carboxamide chloride

Specifications

White-Yellow
50mg
C15H22ClN3O2S3
2811
XXDAXBZYUXLDRD-UHFFFAOYNA-N
hydrogen 5-(2-{[3-(tert-butylamino)-2-hydroxypropyl]sulfanyl}-1,3-thiazol-4-yl)thiophene-2-carboxamide chloride
155032
Crystal-Powder at 20°C
Arotinolol Hydrochloride
236°C
68377-91-3
MFCD00941421
5-[2-[[3-(tert-Butylamino)-2-hydroxypropyl]thio]thiazol-4-yl]thiophene-2-carboxamide Hydrochloride
[H+].[Cl-].CC(C)(C)NCC(O)CSC1=NC(=CS1)C1=CC=C(S1)C(N)=O
407.99
407.99
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : XM8301000

TSCA : No

SDS