Please login to your online account to display your discounted pricing

Aroclor Solution Kits (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)


Quantity 9x1mL (100μg/mL in Methanol)
Packaging Ampules

Manufacturer: ultra scientific  RPCK3A

 View more versions of this product

Catalog No. US-RPCK-3A

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 9x1mL (100μg/mL in Methanol)
Concentration or Composition (by Analyte or Components) Each kit contains one 1mL ampule each:
Arochlor 1016
Arochlor 1221
Arochlor 1232
Arochlor 1242
Arochlor 1248
Arochlor 1254
Arochlor 1260
Arochlor 1262
Arochlor 1268
Methods For EPA 500, 600, 8000/ Appendix IX
Chemical Name or Material Aroclor Solution Kits
Packaging Ampules

Specifications

Chemical Name or Material Aroclor Solution Kits