Please login to your online account to display your discounted pricing

Aroclor Solution Kits (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

$374.91 - $374.91

Chemical Identifiers

Chemical Name or Material Aroclor Solution Kits
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-RPCK-3A

 
ultra scientific
RPCK3A
Ampules 9x1mL (100μg/mL in Methanol) Each for $374.91

US-RPCK-1A

 
ultra scientific
RPCK1A
Ampules 9x1mL (100μg/mL in Hexane) Each for $374.91
Description & Specifications

Specifications

Quantity 9x1mL (100μg/mL in Methanol)
Concentration or Composition (by Analyte or Components) Each kit contains one 1mL ampule each:
Arochlor 1016
Arochlor 1221
Arochlor 1232
Arochlor 1242
Arochlor 1248
Arochlor 1254
Arochlor 1260
Arochlor 1262
Arochlor 1268
Methods For EPA 500, 600, 8000/ Appendix IX
Chemical Name or Material Aroclor Solution Kits
Packaging Ampules