missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aroclor 1268 (PCB 1268) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1268 at 100μg/mL in methanol or hexane

$59.03 - $59.03

Chemical Identifiers

CAS 11100-14-4
Chemical Name or Material Aroclor 1268 (PCB 1268)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-PP-3811 Ultra Scientific
PP3811
Ampule 1x1mL (in Hexane) Each for $59.03
Specifications Description & Specifications

Chemical Identifiers

11100-14-4
Aroclor 1268 (PCB 1268)

Specifications

1x1mL (in Hexane)
Ampule
Aroclor 1268 (PCB 1268)
For EPA 500, 600, 8000/ Appendix IX
11100-14-4