Please login to your online account to display your discounted pricing

Aroclor 1260 PCB Contaminant Kit, Ultra Scientific


Packaging Ampules

Manufacturer: ultra scientific  801175000204

 View more versions of this product

Catalog No. US-1260TK-B

  • / Each
 

Description & Specifications

Chemical Identifiers

Packaging Ampules
Quantity 4 x 1mL
Chemical Name or Material Aroclor 1260 PCB Contaminant Kit
Physical Form Oily liquid

Specifications

Chemical Name or Material Aroclor 1260 PCB Contaminant Kit
Physical Form Oily liquid