Please login to your online account to display your discounted pricing

Aroclor 1260 PCB Contaminant Kit, Ultra Scientific

$285.69 - $285.69

Chemical Identifiers

Chemical Name or Material Aroclor 1260 PCB Contaminant Kit
Physical Form Oily liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-1260TK-B

 
ultra scientific
801175000204
Ampules Each for $285.69
Description & Specifications

Specifications

Packaging Ampules
Quantity 4 x 1mL
Chemical Name or Material Aroclor 1260 PCB Contaminant Kit
Physical Form Oily liquid