missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aroclor 1260 (PCB 1260) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1260 at 100μg/mL in methanol or hexane

$56.74 - $237.46

Chemical Identifiers

CAS 11096-82-5
Chemical Name or Material Aroclor 1260 (PCB 1260)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-PP-3601 Ultra Scientific
PP3601
Ampule 1x1mL (in Methanol) Each for $56.74
US-PP-3611 Ultra Scientific
PP3611
Ampule 1x1mL (in Hexane) Each for $56.74
US-RPC-1260 Ultra Scientific
RPC1260
Glass Vial 50mg (Neat) Each for $237.46
Specifications Description & Specifications

Chemical Identifiers

11096-82-5
Aroclor 1260 (PCB 1260)

Specifications

1x1mL (in Methanol)
11096-82-5
Aroclor 1260 (PCB 1260)
For EPA 500, 600, 8000/ Appendix IX
Ampule