missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aroclor 1254 (PCB 1254) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1254 at 100μg/mL in methanol or hexane

$57.63 - $241.20

Chemical Identifiers

CAS 11097-69-1
Chemical Name or Material Aroclor 1254 (PCB 1254)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-PP-3501 Ultra Scientific
PP3501
Ampule 1x1mL (in Methanol) Each for $57.63
US-PP-3511 Ultra Scientific
PP3511
Ampule 1x1mL (in Hexane) Each for $57.63
US-RPC-1254 Ultra Scientific
RPC1254
Glass Vial 50mg (Neat) Each for $241.20
Specifications Description & Specifications

Chemical Identifiers

11097-69-1
Aroclor 1254 (PCB 1254)

Specifications

1x1mL (in Methanol)
Ampule
Aroclor 1254 (PCB 1254)
For EPA 500, 600, 8000/ Appendix IX
11097-69-1