Please login to your online account to display your discounted pricing

Aroclor 1254 (PCB 1254) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)


Aroclor 1254 at 100μg/mL in methanol or hexane

Quantity 50mg (Neat)
Packaging Glass Vial

Manufacturer: ultra scientific  RPC1254

 View more versions of this product

Catalog No. US-RPC-1254

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg (Neat)
Methods For EPA 500, 600, 8000/ Appendix IX
Chemical Name or Material Aroclor 1254 (PCB 1254)
CAS 11097-69-1
Packaging Glass Vial

Specifications

Chemical Name or Material Aroclor 1254 (PCB 1254)
CAS 11097-69-1