missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aroclor 1248 (PCB 1248) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1248 at 100μg/mL in methanol or hexane

$62.69 - $62.69

Chemical Identifiers

CAS 12672-29-6
Chemical Name or Material Aroclor 1248 (PCB 1248)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-PP-3401 Ultra Scientific
PP3401
Ampule 1x1mL (in Methanol) Each for $62.69
US-PP-3411 Ultra Scientific
PP3411
Ampule 1x1mL (in Hexane) Each for $62.69
Specifications Description & Specifications

Chemical Identifiers

12672-29-6
Aroclor 1248 (PCB 1248)

Specifications

1x1mL (in Methanol)
12672-29-6
Aroclor 1248 (PCB 1248)
For EPA 500, 600, 8000/ Appendix IX
Ampule