Please login to your online account to display your discounted pricing

Aroclor 1242 PCB Contaminant Kit, Ultra Scientific


Quantity 4 x 1mL

Manufacturer: ultra scientific  1242TKB

 View more versions of this product

Catalog No. US-1242TK-B

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Oily Liquid
Quantity 4 x 1mL
Chemical Name or Material Aroclor1242 PCB Contaminant Kit
Packaging Ampules

Specifications

Chemical Name or Material Aroclor1242 PCB Contaminant Kit
Physical Form Oily Liquid