Please login to your online account to display your discounted pricing

Aroclor 1242 PCB Contaminant Kit, Ultra Scientific

$289.77 - $289.77

Chemical Identifiers

Chemical Name or Material Aroclor1242 PCB Contaminant Kit
Physical Form Oily Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-1242TK-B

 
ultra scientific
1242TKB
4 x 1mL Each for $289.77
Description & Specifications

Specifications

Physical Form Oily Liquid
Quantity 4 x 1mL
Chemical Name or Material Aroclor1242 PCB Contaminant Kit
Packaging Ampules