Please login to your online account to display your discounted pricing

Aroclor 1242 (PCB 1242) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1242 at 100μg/mL in methanol or hexane

$57.11 - $239.00

Chemical Identifiers

Chemical Name or Material Aroclor 1242 (PCB 1242)
CAS 53469-21-9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-PP-3101

 
ultra scientific
PP3101
Ampule 1x1mL (in Methanol) Each for $57.11

US-PP-3111

 
ultra scientific
PP3111
Ampule 1x1mL (in Hexane) Each for $57.11

US-RPC-1242

 
ultra scientific
RPC1242
Glass Vial 50mg (Neat) Each for $239.00
Description & Specifications

Specifications

Quantity 1x1mL (in Methanol)
Methods For EPA 500, 600, 8000/ Appendix IX
CAS 53469-21-9
Chemical Name or Material Aroclor 1242 (PCB 1242)
Packaging Ampule