Please login to your online account to display your discounted pricing

Aroclor 1242 (PCB 1242) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)


Aroclor 1242 at 100μg/mL in methanol or hexane

Quantity 50mg (Neat)
Packaging Glass Vial

Manufacturer: ultra scientific  RPC1242

 View more versions of this product

Catalog No. US-RPC-1242

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg (Neat)
Methods For EPA 500, 600, 8000/ Appendix IX
Packaging Glass Vial
Chemical Name or Material Aroclor 1242 (PCB 1242)
CAS 53469-21-9

Specifications

Chemical Name or Material Aroclor 1242 (PCB 1242)
CAS 53469-21-9