Please login to your online account to display your discounted pricing

Aroclor 1232 (PCB 1232) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)


Aroclor 1232 at 100μg/mL in methanol or hexane

Quantity 1x1mL (in Hexane)
Packaging Ampule

Manufacturer: ultra scientific  PP3011

 View more versions of this product

Catalog No. US-PP-3011

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL (in Hexane)
Methods For EPA 500, 600, 8000/Appendix IX
CAS 11141-16-5
Packaging Ampule
Chemical Name or Material Aroclor 1232 (PCB 1232)

Specifications

Chemical Name or Material Aroclor 1232 (PCB 1232)
CAS 11141-16-5