Please login to your online account to display your discounted pricing

Aroclor 1232 (PCB 1232) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1232 at 100μg/mL in methanol or hexane

$62.40 - $62.40

Chemical Identifiers

Chemical Name or Material Aroclor 1232 (PCB 1232)
CAS 11141-16-5
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-PP-3001

 
ultra scientific
PP3001
Ampule 1x1mL (in Methanol) Each for $62.40

US-PP-3011

 
ultra scientific
PP3011
Ampule 1x1mL (in Hexane) Each for $62.40
Description & Specifications

Specifications

Quantity 1x1mL (in Methanol)
Methods For EPA 500, 600, 8000/Appendix IX
Chemical Name or Material Aroclor 1232 (PCB 1232)
CAS 11141-16-5
Packaging Ampule