Please login to your online account to display your discounted pricing

Aroclor 1221 (PCB 1221) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1221 at 100μg/mL in methanol or hexane

$62.00 - $62.00

Chemical Identifiers

Chemical Name or Material Aroclor 1221 (PCB 1221)
CAS 11104-28-2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-PP-2901

 
ultra scientific
PP2901
Ampule 1x1mL Each for $62.00

US-PP-2911

 
ultra scientific
PP2911
Ampule 1x1mL Each for $62.00
Description & Specifications

Specifications

Quantity 1x1mL
Methods For EPA 500, 600, 8000/ Appendix IX
Chemical Name or Material Aroclor 1221 (PCB 1221)
CAS 11104-28-2
Packaging Ampule