missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aroclor 1016 (PCB 1016) (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Aroclor 1016 at 100μg/mL in methanol or hexane

$63.37 - $63.37

Chemical Identifiers

CAS 12674-11-2
Chemical Name or Material Aroclor 1016 (PCB 1016)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-PP-2801 Ultra Scientific
PP2801
Ampule 1x1mL (in Methanol) Each for $63.37
US-PP-2811 Ultra Scientific
PP2811
Ampule 1x1mL (in Hexane) Each for $63.37
Specifications Description & Specifications

Chemical Identifiers

12674-11-2
Aroclor 1016 (PCB 1016)

Specifications

1x1mL (in Methanol)
Ampule
Aroclor 1016 (PCB 1016)
For EPA 500, 600, 8000/ Appendix IX
12674-11-2