Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, Unlabeled, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143315005MG

 View more versions of this product

Catalog No. NBP1433155M

  • / Each
 

Description & Specifications

Specifications

Antigen IgG
Applications Flow Cytometry
Clone 299Arm
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Liquid
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Host Species Armenian Hamster
Isotype IgG
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This 299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antibody.