Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, PE-Cy7, Isotype control, Novus Biologicals™

$252.00 - $373.00

Specifications

Antigen IgG
Primary or Secondary Primary
Host Species Armenian Hamster
Conjugate PE-Cy7
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14294101

 
novus biologicals canada
NBP14294101MG
0.1mg Each for $373.00

NBP14294105

 
novus biologicals canada
NBP142941005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen IgG
Primary or Secondary Primary
Host Species Armenian Hamster
Conjugate PE-Cy7
Applications Flow Cytometry
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.