missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, PE-Cy5, Isotype control, Novus Biologicals™

$252.00 - $362.00

Specifications

Antigen IgG
Primary or Secondary Primary
Host Species Armenian Hamster
Conjugate PE-Cy5
Applications Flow Cytometry
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14307101 Novus Biologicals Canada
NBP14307101MG
0.1mg Each for $362.00
NBP14307105 Novus Biologicals Canada
NBP143071005MG
0.05mg Each for $252.00
Specifications Description & Specifications

Specifications

IgG
Armenian Hamster
Flow Cytometry
Store at 4°C. Do not freeze.
Primary
PE-Cy5
RUO