Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, Biotin, Isotype control, Novus Biologicals™

$214.00 - $719.00

Specifications

Antigen IgG
Primary or Secondary Primary
Host Species Armenian Hamster
Conjugate Biotin
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1435555

 
novus biologicals canada
NBP14355505MG
0.5mg Each for $719.00

NBP1435555M

 
novus biologicals canada
NBP143555005MG
0.05mg Each for $214.00
Description & Specifications

Specifications

Antigen IgG
Primary or Secondary Primary
Host Species Armenian Hamster
Conjugate Biotin
Applications Flow Cytometry
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.