Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, Biotin, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143555005MG

 View more versions of this product

Catalog No. NBP1435555M

  • / Each
 

Description & Specifications

Specifications

Antigen IgG
Format Conjugated
Applications Flow Cytometry
Isotype IgG
Quantity 0.05mg
Primary or Secondary Primary
Clone 299Arm
Conjugate Biotin
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Host Species Armenian Hamster
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity This 299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antibody.
Storage Requirements Store at 4°C. Do not freeze.