Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, Biotin, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14355505MG

 View more versions of this product

Catalog No. NBP1435555

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Applications Flow Cytometry
Primary or Secondary Primary
Format Conjugated
Quantity 0.5mg
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Host Species Armenian Hamster
Conjugate Biotin
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Isotype IgG
Test Specificity This 299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antibody.
Antigen IgG
Clone 299Arm