Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, APC, Isotype control, Novus Biologicals™

$246.00 - $354.00

Specifications

Antigen IgG
Primary or Secondary Primary
Host Species Armenian Hamster
Conjugate APC
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14299905

 
novus biologicals canada
NBP142999005MG
0.05mg Each for $246.00

NBP14299901

 
novus biologicals canada
NBP14299901MG
0.1mg Each for $354.00
Description & Specifications

Specifications

Antigen IgG
Primary or Secondary Primary
Host Species Armenian Hamster
Conjugate APC
Applications Flow Cytometry
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.